Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057766.1 | - | 5 | 675 |
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834171.1 | - | 11 | 1256 |
Gene Symbol: INTS4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027392.2 | 5 | 682 | |||
GenBank mRNA | BC111554.1 | - | 5 | 675 | 40024359 |