Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273585.4 | XP_005273642.1 | 2 | 204 | ||
XM_005273586.4 | XP_005273643.1 | 2 | 207 | |||
XM_005273587.4 | XP_005273644.1 | 1 | 108 | |||
XM_005273588.4 | XP_005273645.1 | 1 | 234 | |||
XM_005273591.4 | XP_005273648.1 | 1 | 234 | |||
XM_011544596.2 | XP_011542898.1 | 2 | 820 | |||
XM_011544597.2 | XP_011542899.1 | 2 | 172 | |||
XM_011544598.2 | XP_011542900.1 | 1 | 274 | |||
XM_017013707.1 | XP_016869196.1 | 1 | 234 | |||
XM_017013710.1 | XP_016869199.1 | 2 | 180 | |||
XM_017013711.1 | XP_016869200.1 | 1 | 102 | |||
XM_017013714.1 | XP_016869203.1 | 1 | 234 | |||
GenBank mRNA | AK094299.1 | - | 2 | 181 | ||
BC035082.1 | - | 1 | 580 |