Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020844.2 | NP_065895.2 | 2 | 342 | ||
XM_005273584.4 | XP_005273641.1 | 2 | 384 | |||
XM_005273585.4 | XP_005273642.1 | 3 | 395 | |||
XM_005273586.4 | XP_005273643.1 | 3 | 398 | |||
XM_005273587.4 | XP_005273644.1 | 2 | 299 | |||
XM_005273588.4 | XP_005273645.1 | 2 | 425 | |||
XM_005273591.4 | XP_005273648.1 | 2 | 425 | |||
XM_011544598.2 | XP_011542900.1 | 2 | 465 | |||
XM_011544600.2 | XP_011542902.1 | 2 | 203 | |||
XM_011544601.2 | XP_011542903.1 | 2 | 326 | |||
XM_017013705.1 | XP_016869194.1 | 2 | 203 | |||
XM_017013706.1 | XP_016869195.1 | 2 | 424 | |||
XM_017013707.1 | XP_016869196.1 | 2 | 425 | |||
XM_017013708.1 | XP_016869197.1 | 2 | 381 | |||
XM_017013709.1 | XP_016869198.1 | 2 | 381 | |||
XM_017013710.1 | XP_016869199.1 | 3 | 371 | |||
XM_017013711.1 | XP_016869200.1 | 2 | 293 | |||
XM_017013712.1 | XP_016869201.1 | 2 | 381 | |||
XM_017013713.1 | XP_016869202.1 | 2 | 381 | |||
XM_017013714.1 | XP_016869203.1 | 2 | 425 | |||
XM_017013715.1 | XP_016869204.1 | 2 | 381 | |||
GenBank mRNA | AK094299.1 | - | 3 | 372 | ||
AK296010.1 | - | 3 | 195 | |||
BC035082.1 | - | 2 | 771 | |||
DA390356.1 | - | 3 | 496 | |||
DC334299.1 | - | 2 | 202 |