Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010895.2 | NP_001010895.1 | 1 | 291 | ||
NM_020207.4 | NP_064592.2 | 1 | 291 | |||
XM_011518641.2 | XP_011516943.1 | 1 | 291 | |||
XM_011518644.2 | XP_011516946.1 | 1 | 277 | |||
XM_011518645.2 | XP_011516947.1 | 1 | 291 | |||
XM_011518646.2 | XP_011516948.1 | 1 | 291 | |||
XM_011518647.1 | XP_011516949.1 | 1 | 291 | |||
XM_017014707.1 | XP_016870196.1 | 1 | 277 | |||
GenBank mRNA | AK308409.1 | - | 1 | 223 | ||
AL389953.1 | - | 1 | 208 | |||
BC140702.1 | - | 1 | 34 | 9021570 | ||
BC144261.1 | - | 1 | 34 | |||
DB153432.1 | - | 1 | 293 |
Gene Symbol: LINC00476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023389.1 | 1 | 66 | |||
NR_023390.1 | 1 | 66 | ||||
GenBank mRNA | AF194971.1 | - | 1 | 65 | ||
AL389954.1 | - | 1 | 66 |