Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003720.1 | 11 | 354 | |||
GenBank mRNA | AY154892.1 | - | 11 | 354 | ||
BC128170.1 | - | 8 | 228 | |||
BC128173.1 | - | 8 | 228 | |||
BC141914.1 | - | 8 | 228 | |||
BC150502.1 | - | 8 | 228 |