Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113434.4 | NP_001106905.1 | 1 | 656 | ||
XM_005256621.3 | XP_005256678.1 | 1 | 649 | |||
XM_006721514.2 | XP_006721577.1 | 1 | 649 | |||
XM_011523822.2 | XP_011522124.1 | 1 | 649 | |||
GenBank mRNA | AK056436.1 | - | 1 | 622 | ||
BC010612.1 | - | 1 | 654 | |||
BC019938.1 | - | 1 | 644 | |||
BC052777.1 | - | 1 | 656 | |||
BC053952.1 | - | 1 | 656 | |||
BC132738.1 | - | 1 | 630 | 40146760 | ||
BC136630.1 | - | 1 | 630 | 9020620 |