Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287395.1 | NP_001274324.1 | 38, 39 | 4744 | ||
XM_011511075.2 | XP_011509377.1 | 38, 39 | 4951 | |||
XM_011511076.2 | XP_011509378.1 | 38, 39 | 4939 | |||
XM_011511077.2 | XP_011509379.1 | 38, 39 | 4921 | |||
XM_011511078.2 | XP_011509380.1 | 38, 39 | 4912 | |||
XM_011511079.2 | XP_011509381.1 | 38, 39 | 4906 | |||
XM_011511080.2 | XP_011509382.1 | 38, 39 | 6422 | |||
XM_011511081.2 | XP_011509383.1 | 37, 38 | 4846 | |||
XM_011511082.2 | XP_011509384.1 | 37, 38 | 4813 | |||
XM_011511083.2 | XP_011509385.1 | 38, 39 | 4951 | |||
XM_011511084.2 | XP_011509386.1 | 37, 38 | 4753 | |||
XM_011511085.2 | XP_011509387.1 | 32, 33 | 4021 | |||
XM_011511086.2 | XP_011509388.1 | 29, 30 | 3676 | |||
GenBank mRNA | BC042133.1 | - | 3, 4 | 571 |