Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252402.2 | NP_001239331.1 | 5 | 731 | ||
NM_001252404.2 | NP_001239333.1 | 4 | 636 | |||
NM_001280561.1 | NP_001267490.1 | 6 | 1335 | |||
NR_047684.2 | 5 | 733 | ||||
GenBank mRNA | AB196634.1 | - | 6 | 1335 | ||
AI478864.1 | - | 1 | 262 | |||
BC127763.1 | - | 4 | 550 | |||
BC127764.1 | - | 4 | 547 | |||
BC141966.1 | - | 5 | 646 | |||
BC150569.1 | - | 4 | 550 | |||
BC150570.1 | - | 4 | 550 | |||
BX648457.1 | - | 2 | 492 |