Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012978.2 | NP_001012996.1 | 3 | 353 | ||
NM_001159560.1 | NP_001153032.1 | 3 | 395 | |||
GenBank mRNA | AY833564.1 | - | 3 | 301 | ||
BC042818.1 | - | 3 | 302 | 5313992 | ||
BC092450.1 | - | 3 | 273 | 30720892 | ||
BC106955.2 | - | 3 | 354 | 40035908 |