Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206920.1 | NP_001193849.1 | 3 | 578 | ||
NM_001306087.1 | NP_001293016.1 | 3 | 581 | |||
XM_011536720.2 | XP_011535022.1 | 5 | 4360 | |||
XM_011536721.2 | XP_011535023.1 | 5 | 4357 | |||
XM_011536723.2 | XP_011535025.1 | 3 | 846 | |||
XM_011536724.2 | XP_011535026.1 | 3 | 1451 | |||
XM_011536725.1 | XP_011535027.1 | 4 | 770 | |||
XM_017021258.1 | XP_016876747.1 | 4 | 766 | |||
XM_017021259.1 | XP_016876748.1 | 5 | 4360 | |||
XM_017021260.1 | XP_016876749.1 | 4 | 773 | |||
XM_017021261.1 | XP_016876750.1 | 3 | 649 | |||
GenBank mRNA | BC101318.1 | - | 3 | 519 | ||
BC101319.1 | - | 3 | 1592 | |||
BC101320.1 | - | 3 | 525 | |||
BC101321.1 | - | 3 | 525 |