Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102386.2 | NP_001095856.1 | 3 | 290 | ||
XM_011516124.2 | XP_011514426.1 | 3 | 784 | |||
GenBank mRNA | BC147016.1 | - | 3 | 290 | 9056944 | |
BC147017.1 | - | 3 | 290 | 9056945 |