Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102386.2 | NP_001095856.1 | 8 | 1007 | ||
GenBank mRNA | BC147016.1 | - | 8 | 1007 | 9056944 | |
BC147017.1 | - | 8 | 1007 | 9056945 |