Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284260.1 | NP_001271189.1 | 3 | 1038 | ||
NM_001284261.1 | NP_001271190.1 | 3 | 539 | |||
NM_001284262.1 | NP_001271191.1 | 3 | 539 | |||
NM_198285.2 | NP_938026.2 | 3 | 1038 | |||
XM_005249989.3 | XP_005250046.1 | 3 | 1035 | |||
XM_005249990.4 | XP_005250047.1 | 3 | 1035 | |||
XM_006715966.2 | XP_006716029.1 | 3 | 1035 | |||
XM_011516144.2 | XP_011514446.1 | 3 | 1035 | |||
XM_011516145.2 | XP_011514447.1 | 3 | 1035 | |||
XM_011516146.2 | XP_011514448.1 | 3 | 1035 | |||
XM_011516147.2 | XP_011514449.1 | 3 | 1035 | |||
XM_011516148.1 | XP_011514450.1 | 3 | 523 | |||
XM_011516150.1 | XP_011514452.1 | 4 | 727 | |||
XM_011516151.2 | XP_011514453.1 | 3 | 1035 | |||
XM_011516152.1 | XP_011514454.1 | 3 | 1035 | |||
GenBank mRNA | AK125347.1 | - | 3 | 488 | ||
AK296049.1 | - | 3 | 631 | |||
BC107125.2 | - | 2 | 435 | 40023951 | ||
BX390499.1 | - | 1 | 145 |