Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256508.1 | NP_001243437.1 | 5 | 469 | ||
NM_198517.3 | NP_940919.1 | 5 | 486 | |||
NR_046266.1 | 4 | 656 | ||||
XM_006718538.3 | XP_006718601.1 | 5 | 745 | |||
XM_006718539.3 | XP_006718602.1 | 5 | 728 | |||
XM_006718540.3 | XP_006718603.1 | 5 | 658 | |||
XM_006718541.3 | XP_006718604.1 | 5 | 753 | |||
XM_006718542.3 | XP_006718605.1 | 5 | 736 | |||
XM_006718543.2 | XP_006718606.1 | 5 | 470 | |||
XM_006718544.3 | XP_006718607.1 | 5 | 664 | |||
XM_011545001.2 | XP_011543303.1 | 4 | 691 | |||
XM_011545002.2 | XP_011543304.1 | 4 | 706 | |||
GenBank mRNA | AK122585.1 | - | 2 | 328 | ||
AK126131.1 | - | 1 | 1734 | |||
BC022202.1 | - | 4 | 430 | |||
BC036873.1 | - | 4 | 441 | 5224063 | ||
BC062999.1 | - | 5 | 458 | 5176854 | ||
CR979868.1 | - | 5 | 486 |