Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015183.2 | NP_055998.1 | 2, 3 | 133 | ||
NM_198828.2 | NP_942123.1 | 4, 5 | 976 | |||
XM_011543382.2 | XP_011541684.1 | 5, 6 | 1032 | |||
XM_017009447.1 | XP_016864936.1 | 5, 6 | 1032 | |||
GenBank mRNA | AY830839.1 | - | 2, 3 | 134 | ||
BC033215.1 | - | 4, 5 | 944 | 5015614 |