Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 5 | 869 | ||
XM_006712884.3 | XP_006712947.1 | 5 | 1293 | |||
XM_006712887.3 | XP_006712950.1 | 7 | 1246 | |||
XM_017005467.1 | XP_016860956.1 | 4 | 865 | |||
XM_017005468.1 | XP_016860957.1 | 4 | 773 | |||
XM_017005469.1 | XP_016860958.1 | 4 | 762 |
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055744.1 | - | 1 | 1320 | ||
DQ786216.1 | - | 3 | 590 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 1371 | |||
GenBank mRNA | AK128231.1 | - | 4 | 1367 | ||
BC070322.1 | - | 4 | 694 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 9 | 1478 | ||
XM_017015367.1 | XP_016870856.1 | 8 | 1395 | |||
XM_017015368.1 | XP_016870857.1 | 8 | 1378 | |||
XM_017015369.1 | XP_016870858.1 | 9 | 1461 | |||
XM_017015370.1 | XP_016870859.1 | 5 | 1478 | |||
XM_017015371.1 | XP_016870860.1 | 7 | 1245 | |||
XM_017015373.1 | XP_016870862.1 | 6 | 1116 | |||
XM_017015374.1 | XP_016870863.1 | 6 | 1054 | |||
XM_017015375.1 | XP_016870864.1 | 5 | 970 |