Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099666.1 | NP_001093136.1 | 6 | 898 | ||
XM_005251976.4 | XP_005252033.1 | 6 | 988 | |||
XM_005251977.4 | XP_005252034.1 | 6 | 962 | |||
XM_011518639.1 | XP_011516941.1 | 6 | 862 | |||
XM_011518640.1 | XP_011516942.1 | 7 | 1038 | |||
GenBank mRNA | AK127511.1 | - | 1 | 152 | ||
AL832683.1 | - | 4 | 642 | |||
BC053622.1 | - | 6 | 880 |