Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519443.1 | XP_011517745.1 | 8, 9 | 651 | ||
XM_017016029.1 | XP_016871518.1 | 12, 13 | 1792 | |||
XM_017016030.1 | XP_016871519.1 | 11, 12 | 1194 | |||
XM_017016031.1 | XP_016871520.1 | 11, 12 | 1205 |