Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142769.1 | NP_001136241.1 | 35 | 4872 | ||
NM_001142770.1 | NP_001136242.1 | 34 | 4907 | |||
NM_001142771.1 | NP_001136243.1 | 34 | 4851 | |||
NM_001142772.1 | NP_001136244.1 | 33 | 4836 | |||
XM_017016571.1 | XP_016872060.1 | 36 | 5557 | |||
XM_017016572.1 | XP_016872061.1 | 33 | 5426 | |||
XM_017016573.1 | XP_016872062.1 | 40 | 8233 | |||
GenBank mRNA | EU718480.1 | - | 34 | 4851 | ||
EU718481.1 | - | 34 | 4913 | |||
EU718482.1 | - | 33 | 4830 |