Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012754.3 | NP_001012772.1 | 3 | 664 | ||
NM_001017370.2 | NP_001017370.1 | 3 | 664 | |||
NR_073109.1 | 3 | 406 | ||||
GenBank mRNA | AK299198.1 | - | 3 | 577 | ||
AL833329.1 | - | 2 | 1081 | |||
BC052492.1 | - | 3 | 344 | |||
BC140877.1 | - | 3 | 590 | 9021745 | ||
BC140878.1 | - | 3 | 590 | 9021746 | ||
BX647872.1 | - | 3 | 554 | |||
BX648122.1 | - | 3 | 616 | |||
CR749318.1 | - | 3 | 585 | |||
DA772703.1 | - | 3 | 406 |