Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207380.2 | NP_997263.2 | 11 | 1415 | ||
GenBank mRNA | AK125329.1 | - | 11 | 1408 | ||
AK126035.1 | - | 7 | 805 | |||
BC140897.1 | - | 7 | 804 | 9021765 | ||
BC140902.1 | - | 7 | 804 | 9021770 | ||
CR749408.1 | - | 8 | 1109 |