Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720543.3 | XP_006720606.1 | 17 | 4741 | ||
XM_006720544.3 | XP_006720607.1 | 16 | 4717 | |||
XM_017022240.1 | XP_016877729.1 | 18 | 4612 | |||
XM_017022241.1 | XP_016877730.1 | 17 | 4097 | |||
XM_017022243.1 | XP_016877732.1 | 17 | 4093 | |||
XM_017022244.1 | XP_016877733.1 | 17 | 3995 | |||
XM_017022245.1 | XP_016877734.1 | 18 | 4050 | |||
XM_017022246.1 | XP_016877735.1 | 17 | 4151 | |||
XM_017022251.1 | XP_016877740.1 | 10 | 3112 | |||
XM_017022252.1 | XP_016877741.1 | 9 | 3088 | |||
XM_017022253.1 | XP_016877742.1 | 18 | 4783 | |||
XM_017022254.1 | XP_016877743.1 | 18 | 4816 |