siRNA Details
Gene Symbol: BHMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310124.1 | NP_001297053.1 | 3 | 572 | ||
XM_011527571.2 | XP_011525873.1 | 3 | 184 | |||
XM_011527573.2 | XP_011525875.1 | 2 | 352 | |||
GenBank mRNA | BI463439.1 | - | 3 | 583 |
