Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004807.2 | NP_004798.3 | 2 | 970 | ||
GenBank mRNA | AB006179.1 | - | 2 | 1044 | ||
AK293724.1 | - | 2 | 427 | |||
AK295898.1 | - | 2 | 812 | |||
BC001196.1 | - | 1 | 219 | |||
BC096239.1 | - | 2 | 946 | 40005544 | ||
BC096240.1 | - | 2 | 946 | 40005547 | ||
BC099638.3 | - | 2 | 946 | 40005546 | ||
BC099639.1 | - | 2 | 946 | 40005548 |