Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010978.3 | NP_001010978.2 | 3 | 240 | ||
XM_011541446.1 | XP_011539748.1 | 3 | 220 | |||
GenBank mRNA | BC127708.1 | - | 3 | 237 | 40127476 | |
CF887342.1 | - | 3 | 240 |