Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100624.2 | NP_001094094.2 | 11 | 3935 | ||
NM_001270473.1 | NP_001257402.1 | 10 | 3875 | |||
NM_001270474.1 | NP_001257403.1 | 10 | 3833 | |||
XM_006721236.3 | XP_006721299.1 | 11 | 4084 |
Gene Symbol: GATS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124689.1 | - | 2 | 3757 |
Gene Symbol: HEATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182922.2 | NP_891552.1 | 15 | 2358 | ||
XM_005256013.2 | XP_005256070.1 | 14 | 2173 | |||
XM_011523188.2 | XP_011521490.1 | 15 | 2248 | |||
XM_017023385.1 | XP_016878874.1 | 13 | 2311 | |||
XM_017023386.1 | XP_016878875.1 | 13 | 2116 |
Gene Symbol: LOC105371242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510267.2 | XP_011508569.1 | 1 | 2519 |
Gene Symbol: PPIAL4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123006.1 | - | 2 | 2753 |
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094769.1 | - | 1 | 252 |