Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530438.2 | XP_011528740.1 | 1 | 215 | ||
XM_017028990.1 | XP_016884479.1 | 1 | 215 |