Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080473.1 | NP_001073942.1 | 13 | 1418 | ||
XM_005264301.4 | XP_005264358.1 | 13 | 2479 | |||
XM_017004063.1 | XP_016859552.1 | 12 | 2572 | |||
XM_017004064.1 | XP_016859553.1 | 12 | 2557 | |||
XM_017004065.1 | XP_016859554.1 | 11 | 2488 | |||
GenBank mRNA | BX647146.1 | - | 7 | 1183 |