Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035505.1 | NP_001030582.1 | 3 | 317 | ||
NM_212552.2 | NP_997717.2 | 4 | 406 | |||
GenBank mRNA | AI652095.1 | - | 1 | 143 | ||
BC017744.2 | - | 2 | 242 | |||
BC028282.1 | - | 3 | 277 | |||
BC042036.1 | - | 4 | 402 | |||
BU600037.1 | - | 411 |