Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085420.1 | NP_001078889.1 | 3, 4 | 1227 | ||
NM_001321245.1 | NP_001308174.1 | 3, 4 | 1191 | |||
XM_017006373.1 | XP_016861862.1 | 3, 4 | 438 | |||
GenBank mRNA | AY436642.1 | - | 3, 4 | 1227 | ||
BC157886.1 | - | 3, 4 | 214 | 8862707 | ||
BC171917.1 | - | 3, 4 | 214 | 9054571 |