Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152617.3 | NP_689830.2 | 6 | 3426 | ||
GenBank mRNA | AK097068.1 | - | 1 | 308 |