Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205854.2 | NP_995326.1 | 1 | 123 | ||
GenBank mRNA | AY102070.1 | - | 1 | 98 | ||
AY359057.1 | - | 1 | 101 | |||
BC039677.1 | - | 1 | 123 | 5186107 |