Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125823.1 | 3 | 543 | |||
GenBank mRNA | AI025346.1 | - | 1 | 115 | ||
BX094193.1 | - | 3 | 315 |