Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011720.1 | NP_001011720.1 | 4 | 975 | ||
NM_001287258.1 | NP_001274187.1 | 5 | 1093 | |||
NM_001287259.1 | NP_001274188.1 | 5 | 1085 | |||
NM_001287260.1 | NP_001274189.1 | 4 | 829 | |||
XM_006716447.3 | XP_006716510.1 | 2 | 325 | |||
XM_011517524.2 | XP_011515826.1 | 4 | 958 | |||
XM_011517525.2 | XP_011515827.1 | 4 | 820 | |||
XM_011517526.2 | XP_011515828.1 | 4 | 970 | |||
XM_011517527.2 | XP_011515829.1 | 4 | 970 | |||
XM_017013404.1 | XP_016868893.1 | 3 | 1192 | |||
XM_017013405.1 | XP_016868894.1 | 4 | 970 | |||
GenBank mRNA | AY534247.1 | - | 4 | 975 | ||
BC137104.1 | - | 4 | 893 | 9021101 | ||
BC137105.1 | - | 4 | 829 | 9021102 | ||
BC151845.1 | - | 5 | 1052 |