Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137601.2 | NP_001131073.1 | 1 | 82 | ||
XM_017020911.1 | XP_016876400.1 | 2 | 186 | |||
GenBank mRNA | AX721091.1 | - | 2 | 186 | ||
BC157833.1 | - | 1 | 92 | 9057052 | ||
BC171822.1 | - | 1 | 92 | 9054476 |