Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSL3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027917.1 | 4, 5 | 508 | |||
GenBank mRNA | AJ851862.1 | - | 4, 5 | 508 |