Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLOD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080489.2 | NP_001073958.2 | 3 | 294 | ||
XM_011543911.2 | XP_011542213.1 | 4 | 397 | |||
XM_011543912.2 | XP_011542214.1 | 4 | 541 | |||
GenBank mRNA | DY655159.1 | - | 2 | 135 |