Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207645.3 | NP_997528.2 | 2 | 583 | ||
XM_011542817.2 | XP_011541119.1 | 2 | 687 | |||
XM_011542818.2 | XP_011541120.1 | 2 | 565 | |||
GenBank mRNA | AK303693.1 | - | 2 | 583 | ||
BC035798.1 | - | 2 | 503 | 5735100 | ||
BC068577.1 | - | 2 | 556 | 4818568 |