Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 1, 2 | 283 | |||
NR_038344.1 | 1, 2 | 283 | ||||
GenBank mRNA | AK311103.1 | - | 1, 2 | 251 | ||
BI521274.1 | - | 1, 2 | 283 | |||
CN311982.1 | - | 1, 2 | 250 |