Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004737.1 | NP_001004737.1 | 1 | 802 | ||
GenBank mRNA | BC136724.1 | - | 1 | 879 | 9020714 | |
BC136725.1 | - | 1 | 879 | 9020715 |