Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098175.1 | NP_001091645.1 | 10 | 10911 | ||
NM_001164178.1 | NP_001157650.1 | 10 | 10811 | |||
NM_001164179.1 | NP_001157651.1 | 10 | 10913 | |||
NM_001164181.1 | NP_001157653.1 | 9 | 10683 | |||
NM_001164182.1 | NP_001157654.1 | 9 | 10885 | |||
NM_001164183.1 | NP_001157655.1 | 8 | 10767 | |||
NM_001312654.1 | NP_001299583.1 | 10 | 10857 | |||
NM_001776.5 | NP_001767.3 | 10 | 11036 | |||
XM_011540370.2 | XP_011538672.2 | 9 | 10771 | |||
XM_011540371.2 | XP_011538673.1 | 11 | 11047 | |||
XM_011540373.2 | XP_011538675.2 | 9 | 10834 | |||
XM_011540377.2 | XP_011538679.1 | 10 | 10918 | |||
XM_017016959.1 | XP_016872448.1 | 8 | 10501 | |||
GenBank mRNA | AK098240.1 | - | 1 | 1208 | ||
AL833543.1 | - | 1 | 4719 | |||
BX537522.1 | - | 1 | 7175 |