Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083914.2 | NP_001077383.1 | 1 | 94 | ||
XM_005269561.2 | XP_005269618.1 | 1 | 63 | |||
XM_005269568.4 | XP_005269625.1 | 1 | 71 | |||
XM_005269569.2 | XP_005269626.1 | 1 | 63 | |||
XM_005269572.3 | XP_005269629.1 | 1 | 72 | |||
XM_006717642.2 | XP_006717705.1 | 1 | 63 | |||
XM_011539351.1 | XP_011537653.1 | 1 | 63 | |||
XM_011539353.1 | XP_011537655.1 | 1 | 63 | |||
XM_011539354.1 | XP_011537656.1 | 1 | 63 | |||
XM_011539357.1 | XP_011537659.1 | 1 | 63 | |||
GenBank mRNA | BC047018.1 | - | 1 | 59 | ||
CN353451.1 | - | 1 | 90 | |||
DB456766.1 | - | 1 | 94 |