Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014620.5 | NP_055435.2 | 4 | 2143 | ||
NM_153633.2 | NP_705897.1 | 2 | 1503 | |||
GenBank mRNA | AA331371.1 | - | 1 | 63 | ||
BC050442.1 | - | 2 | 1499 | 6186995 | ||
CA432146.1 | - | 1 | 167 |