Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719868.3 | XP_006719931.2 | 1 | 20 |
Gene Symbol: MZT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001071775.2 | NP_001065243.1 | 1 | 39 | ||
GenBank mRNA | BC125183.1 | - | 1 | 42 | 40122127 |