Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135827.1 | 1 | 76 | |||
NR_135828.1 | 1 | 76 | ||||
NR_135831.1 | 1 | 76 | ||||
GenBank mRNA | AK126286.1 | - | 1 | 76 |
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522014.2 | XP_011520316.1 | 1 | 188 | ||
XM_011522019.2 | XP_011520321.1 | 1 | 188 |