Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIPT-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015396.1 | 5 | 545 | |||
NR_024370.1 | 5 | 422 | ||||
GenBank mRNA | AI269516.1 | - | 2 | 234 | ||
AK131031.1 | - | 5 | 545 | |||
BC070380.1 | - | 5 | 527 | |||
BM006535.1 | - | 5 | 423 |