Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014501.2 | NP_055316.2 | 4 | 959 | ||
GenBank mRNA | AB062397.1 | - | 1 | 769 | ||
BC004236.2 | - | 1 | 769 | 2988449 | ||
BC007554.1 | - | 1 | 769 | 2967547 | ||
BC065364.1 | - | 1 | 779 | 5434942 | ||
BC066948.1 | - | 1 | 615 | 4836851 | ||
CA421746.1 | - | 1 | 245 | |||
M91670.1 | - | 4 | 650 |