Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000674.2 | NP_000665.1 | 1 | 178 | ||
NM_001048230.1 | NP_001041695.1 | 1 | 178 | |||
XM_005244901.1 | XP_005244958.1 | 1 | 181 | |||
XM_017000342.1 | XP_016855831.1 | 1 | 135 | |||
GenBank mRNA | AK095060.1 | - | 1 | 163 | ||
AK127752.1 | - | 1 | 821 | |||
AK289928.1 | - | 1 | 178 | |||
BC026340.1 | - | 1 | 156 | 4797116 | ||
DA343445.1 | - | 1 | 178 | |||
DB171678.1 | - | 1 | 181 | |||
L22214.1 | - | 3 | 156 | |||
X68485.1 | - | 1 | 337 |
Set Membership: |
G-protein coupled receptor |