Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166242.1 | NP_001159714.1 | 1 | 78 | ||
NM_173793.4 | NP_776154.3 | 1 | 78 | |||
GenBank mRNA | AK290073.1 | - | 1 | 51 | ||
BX337505.2 | - | 1 | 78 |