Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264633.2 | XP_005264690.1 | 1 | 517 | ||
XM_005264634.2 | XP_005264691.1 | 1 | 517 | |||
XM_011533148.2 | XP_011531450.1 | 1 | 517 | |||
XM_011533149.2 | XP_011531451.1 | 1 | 517 | |||
XM_011533150.2 | XP_011531452.1 | 1 | 517 | |||
XM_011533151.2 | XP_011531453.1 | 1 | 517 | |||
XM_011533152.2 | XP_011531454.1 | 1 | 517 | |||
XM_011533153.2 | XP_011531455.1 | 1 | 517 | |||
XM_017005226.1 | XP_016860715.1 | 1 | 517 |
Gene Symbol: LOC440862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123751.1 | - | 1 | 101 |